Chlorotrihexylsilane ≥95.0% (by GC)

Supplier: TCI America
Danger

C3556-25ML C3556-5ML
TCC3556-25MLEA 196 USD
TCC3556-25ML TCC3556-5ML
Chlorotrihexylsilane ≥95.0% (by GC)
Chlorotrihexylsilane
Formula: C₁₈H₃₉ClSi
MW: 319.05 g/mol
Boiling Pt: 154 °C
MDL Number: MFCD00000510
CAS Number: 3634-67-1
EINECS: 222-851-9
UN: 2987
ADR: 8,II

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR