4-Chloro-3-fluoropyridine ≥95.0% (by GC)

Supplier: TCI America
Warning

C3245-1G C3245-5G
TCC3245-1GEA 62 USD
TCC3245-1G TCC3245-5G
4-Chloro-3-fluoropyridine ≥95.0% (by GC)
4-Chloro-3-fluoropyridine
Formula: C₅H₃ClFN
Boiling Pt: 139 °C
Melting Pt: –24 °C
Flash Pt: 28 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03453233
CAS Number: 2546-56-7
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR