4-Chloro-1-butene ≥98.0% (by GC)

Supplier: TCI America
Danger

Synonyms: 4-Chlorobut-1-ene

C3611-25G C3611-5G
TCC3611-25GEA 183 USD
TCC3611-25G TCC3611-5G
4-Chloro-1-butene ≥98.0% (by GC)
4-Chloro-1-butene
Formula: C₄H₇Cl
MW: 90.55 g/mol
Boiling Pt: 75 °C
MDL Number: MFCD00039386
CAS Number: 927-73-1
EINECS: 213-160-3
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR