3-Chloro-5-fluorobenzaldehyde ≥98.0% (by GC)

Supplier: TCI America
Warning

C3365-1G C3365-5G
TCC3365-1GEA 62 USD
TCC3365-1G TCC3365-5G
3-Chloro-5-fluorobenzaldehyde ≥98.0% (by GC)
3-Chloro-5-fluorobenzaldehyde
Formula: C₇H₄ClFO
MW: 158.56 g/mol
MDL Number: MFCD02261734
CAS Number: 90390-49-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 40.0 to 44.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR