3-Chloro-p-cresol ≥98.0% (by GC)

Supplier: TCI America
Danger

Synonyms: 3-Chloro-4-methylphenol, 1-Chloro-3-hydroxy-6-methylbenzene

C3333-25G C3333-5G
TCC3333-25GEA 206 USD
TCC3333-25G TCC3333-5G
3-Chloro-p-cresol ≥98.0% (by GC)
3-Chloro-p-cresol
Formula: C₇H₇ClO
Boiling Pt: 228 °C
MDL Number: MFCD00060319
CAS Number: 615-62-3
EINECS: 210-439-1
UN: 3437
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Light yellow to Amber to Dark green powder to lump
Purity (GC) min. 98.0 %
Melting point 51.0 to 55.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR