4-Bromo-2,6-dichlorophenol ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
B5034-25G
B5034-5G
TCB5034-25GEA
604
USD
TCB5034-25G
TCB5034-5G
4-Bromo-2,6-dichlorophenol ≥98.0% (by GC, titration analysis)
4-Bromo-2,6-dichlorophenol
Formula:
C₆H₃BrCl₂O Boiling Pt: 266 °C |
MDL Number:
MFCD00019996 CAS Number: 3217-15-0 EINECS: 221-740-2 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | gt. 62.0 and le 66.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...