α-Bromo-4-(difluoromethoxy)toluene ≥98.0% (by GC)

Supplier: TCI America
Danger

Synonyms: 1-(Bromomethyl)-4-(difluoromethoxy)benzene, p-(Difluoromethoxy)benzyl bromide, 4-(Difluoromethoxy)benzyl bromide

D4956-1G D4956-5G
TCD4956-1GEA 71 USD
TCD4956-1G TCD4956-5G
α-Bromo-4-(difluoromethoxy)toluene ≥98.0% (by GC)
α-Bromo-4-(difluoromethoxy)toluene
Formula: C₈H₇BrF₂O
Boiling Pt: 52 °C
MDL Number: MFCD00085007
CAS Number: 3447-53-8
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR