2,3-Difluorobenzylamine ≥98.0% (by GC)

Supplier: TCI America
Danger

Synonyms: (2,3-Difluorophenyl)methanamine

D5133-1G D5133-5G
TCD5133-1GEA 164 USD
TCD5133-1G TCD5133-5G
2,3-Difluorobenzylamine ≥98.0% (by GC)
2,3-Difluorobenzylamine
Formula: C₇H₇F₂N
MW: 143.14 g/mol
Flash Pt: 77 °C
MDL Number: MFCD00070793
CAS Number: 72235-51-9
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR