2,4-Xylyl isothiocyanate ≥98.0% (by GC)

Supplier: TCI America
Danger

Synonyms: 2,4-Dimethylphenylisothiocyanate, 2,4-Dimethylphenyl isothiocyanate

D4913-25G D4913-5G
TCD4913-25GEA 389 USD
TCD4913-25G TCD4913-5G
2,4-Xylyl isothiocyanate ≥98.0% (by GC)
2,4-Xylyl isothiocyanate
Formula: C₉H₉NS
MW: 163.24 g/mol
Boiling Pt: 263 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00022054
CAS Number: 39842-01-8
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White or Colorless to Light orange to Yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR