(2,4-Bis(trifluoromethyl)phenyl)acetic acid ≥97.0% (by titrimetric analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
2,4-Bis(trifluoromethyl)phenylacetic acid
B5337-1G
B5337-5G
TCB5337-1GEA
79
USD
TCB5337-1G
TCB5337-5G
(2,4-Bis(trifluoromethyl)phenyl)acetic acid ≥97.0% (by titrimetric analysis)
(2,4-Bis(trifluoromethyl)phenyl)acetic acid
Formula:
(C₆H₃(CF₃)₂)OCH₂COOH MW: 272.15 g/mol |
MDL Number:
MFCD00042492 CAS Number: 177952-39-5 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 96.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Melting point | 83.0 to 87.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...