2-Bromo-5-methoxypyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

B5910-1G B5910-5G
TCB5910-1GEA 35 USD
TCB5910-1G TCB5910-5G
2-Bromo-5-methoxypyridine ≥98.0% (by GC, titration analysis)
2-Bromo-5-methoxypyridine
Formula: C₆H₆BrNO
Boiling Pt: 78 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07781221
CAS Number: 105170-27-2

Order Now

Specification Test Results

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR