2,4-Dibromo-6-nitrophenol ≥98.0% (by GC)

Supplier: TCI America
Warning

D5323-1G D5323-5G
TCD5323-1GEA 109 USD
TCD5323-1G TCD5323-5G
2,4-Dibromo-6-nitrophenol ≥98.0% (by GC)
2,4-Dibromo-6-nitrophenol
Formula: C₆H₃Br₂NO₃
MDL Number: MFCD00024232
CAS Number: 15969-09-2

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 116.0 to 120.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR