Copper(I) thiocyanate ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

C3417-250G C3417-25G
TCC3417-250GEA 96 USD
TCC3417-250G TCC3417-25G
Copper(I) thiocyanate ≥98.0% (by titrimetric analysis)
Copper(I) thiocyanate
Formula: CCuNS
MDL Number: MFCD00010980
CAS Number: 1111-67-7
EINECS: 214-183-1
UN: 3077
ADR: 9,III
Merck Index: 14,02669

Order Now

Specification Test Results

Appearance White to Gray to Red powder to crystal
Purity (Chelometric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR