2-Chloro-6-nitrobenzaldehyde ≥98.0% (by GC)

Supplier: TCI America
Warning

C3593-1G C3593-5G
TCC3593-1GEA 70 USD
TCC3593-1G TCC3593-5G
2-Chloro-6-nitrobenzaldehyde ≥98.0% (by GC)
2-Chloro-6-nitrobenzaldehyde
Formula: C₇H₄ClNO₃
MDL Number: MFCD00007204
CAS Number: 6361-22-4
EINECS: 228-831-6

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 68.0 to 72.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR