2-Amino-3-bromo-5-fluoropyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

A3346-1G A3346-5G
TCA3346-1GEA 62 USD
TCA3346-1G TCA3346-5G
2-Amino-3-bromo-5-fluoropyridine ≥98.0% (by GC, titration analysis)
2-Amino-3-bromo-5-fluoropyridine
Formula: C₅H₄BrFN₂
MDL Number: MFCD03092924
CAS Number: 869557-43-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 64.0 to 68.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR