Adamantane ≥99.5% (by GC), purified by sublimation

Supplier: TCI America
Warning

Synonyms: Tricyclo[3.3.1.1(3,7)]decane

A3233-25G A3233-5G
TCA3233-25GEA 288 USD
TCA3233-25G TCA3233-5G
Adamantane ≥99.5% (by GC), purified by sublimation
Adamantane
Formula: C₁₀H₁₆
MW: 136.24 g/mol
MDL Number: MFCD00074719
CAS Number: 281-23-2
EINECS: 206-001-4
Merck Index: 14,00149

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR