1-Bromo-2-methoxynaphthalene ≥98.0% (by GC)

Supplier: TCI America
Warning

B6103-25G B6103-5G
TCB6103-25GEA 189 USD
TCB6103-25G TCB6103-5G
1-Bromo-2-methoxynaphthalene ≥98.0% (by GC)
1-Bromo-2-methoxynaphthalene
Formula: C₁₁H₉BrO
Boiling Pt: 192 °C
MDL Number: MFCD00055374
CAS Number: 3401-47-6
EINECS: 222-272-1

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 84.0 to 88.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR