4-Decylaniline ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

D2731-25G D2731-5G
TCD2731-25GEA 416 USD
TCD2731-25G TCD2731-5G
4-Decylaniline ≥98.0% (by GC, titration analysis)
4-Decylaniline
Formula: C₁₆H₂₇N
MW: 233.4 g/mol
Boiling Pt: 164 °C
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007918
CAS Number: 37529-30-9
EINECS: 253-546-9
UN: 3082
ADR: 9 III

Order Now

Specification Test Results

Appearance Colorless to Brown clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR