4-Chloro-2-fluoro-5-nitrotoluene ≥98.0% (by GC)

Supplier: TCI America
Warning

C3334-25G C3334-5G
TCC3334-25GEA 365 USD
TCC3334-25G TCC3334-5G
4-Chloro-2-fluoro-5-nitrotoluene ≥98.0% (by GC)
4-Chloro-2-fluoro-5-nitrotoluene
Formula: C₇H₅ClFNO₂
MDL Number: MFCD00134231
CAS Number: 18349-11-6
EINECS: 242-225-9

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 41.0 to 45.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR