1-Chloro-4-ethylbenzene ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 4-Chloro(ethylbenzene)

C3091-25G C3091-5G
TCC3091-25GEA 464 USD
TCC3091-25G TCC3091-5G
1-Chloro-4-ethylbenzene ≥98.0% (by GC)
1-Chloro-4-ethylbenzene
Formula: C₈H₉Cl
MW: 140.61 g/mol
Boiling Pt: 184 °C
Flash Pt: 60 °C
MDL Number: MFCD00040855
CAS Number: 622-98-0
EINECS: 210-763-3
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR