3,5-Dinitrosalicylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

Synonyms: 2-Hydroxy-3,5-dinitrobenzoic acid, 3,5-Dinitro-2-hydroxybenzoic acid

D0850-100G D0850-25G D0850-500G
TCD0850-100GEA 97 USD
TCD0850-100G TCD0850-25G TCD0850-500G
3,5-Dinitrosalicylic acid ≥98.0% (by GC, titration analysis)
3,5-Dinitrosalicylic acid
Formula: C₇H₄N₂O₇
MDL Number: MFCD00007104
CAS Number: 609-99-4
EINECS: 210-204-3
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 172.0 to 175.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR