2,4-Dimethoxybenzonitrile ≥99.0% (by GC)

Supplier: TCI America
Danger

D4854-25G D4854-5G
TCD4854-25GEA 158 USD
TCD4854-25G TCD4854-5G
2,4-Dimethoxybenzonitrile ≥99.0% (by GC)
2,4-Dimethoxybenzonitrile
Formula: C₉H₉NO₂
MW: 163.18 g/mol
MDL Number: MFCD00001786
CAS Number: 4107-65-7
EINECS: 223-886-2
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 99.0 %
Melting point 93.0 to 97.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR