1-Bromo-3-nitrobenzene ≥98.0% (by GC)

Supplier: TCI America
Danger

B0623-25G B0623-500G B0623-100G
TCB0623-25GEA 25 USD
TCB0623-25G TCB0623-500G TCB0623-100G
1-Bromo-3-nitrobenzene ≥98.0% (by GC)
1-Bromo-3-nitrobenzene
Formula: C₆H₄BrNO₂
MW: 202.01 g/mol
Flash Pt: (closed cup)
MDL Number: MFCD00024298
CAS Number: 585-79-5
EINECS: 209-563-9
UN: 3459
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 54.0 to 56.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR