2,6-Dibromoaniline ≥98.0% (by GC)

Supplier: TCI America
Warning

D0004-25G D0004-5G
TCD0004-25GEA 217 USD
TCD0004-25G TCD0004-5G
2,6-Dibromoaniline ≥98.0% (by GC)
2,6-Dibromoaniline
Formula: C₆H₅Br₂N
MW: 250.92
Boiling Pt: 262
MDL Number: MFCD00007638
CAS Number: 608-30-0
EINECS: 210-158-4

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 81.0 to 84.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR