1,2-Dibromotetrafluorobenzene ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 1,2-Dibromo-3,4,5,6-tetrafluorobenzene

D1479-1G D1479-5G
TCD1479-1GEA 44 USD
TCD1479-1G TCD1479-5G
1,2-Dibromotetrafluorobenzene ≥98.0% (by GC)
1,2-Dibromotetrafluorobenzene
Formula: C₆Br₂F₄
MW: 307.87 g/mol
Boiling Pt: 96 °C
Melting Pt: 12
Flash Pt: 99 °C
MDL Number: MFCD00039209
CAS Number: 827-08-7
EINECS: 212-564-7

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR