1-Bromo-4-chloro-2-nitrobenzene ≥98.0% (by GC)

Supplier: TCI America
Warning

B5696-25G B5696-5G
TCB5696-25GEA 111 USD
TCB5696-25G TCB5696-5G
1-Bromo-4-chloro-2-nitrobenzene ≥98.0% (by GC)
1-Bromo-4-chloro-2-nitrobenzene
Formula: C₆H₃BrClNO₂
MDL Number: MFCD00024320
CAS Number: 41513-04-6
EINECS: 255-421-4

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 67.0 to 71.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR