1-Chloro-2-propanol ≥98.0% (by GC)

Supplier: TCI America
Danger

C3533-1G C3533-5G
TCC3533-1GEA 83 USD
TCC3533-1G TCC3533-5G
1-Chloro-2-propanol ≥98.0% (by GC)
1-Chloro-2-propanol
Formula: C₃H₇ClO
Boiling Pt: 132 °C
Flash Pt: 52 °C
MDL Number: MFCD00004530
CAS Number: 127-00-4
UN: 2611
ADR: 6.1,II
Merck Index: 14,07851

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR