1,1-Dibromo-3,3,3-trifluoroacetone ≥98.0% (by GC)

Supplier: TCI America
Danger

Synonyms: 3,3-Dibromo-1,1,1-trifluoroacetone, 3,3-Dibromo-1,1,1-trifluoro-2-propanone, 3,3-Dibromo-1,1,1-trifluoropropan-2-one

D4004-25G D4004-5G
TCD4004-25GEA 149 USD
TCD4004-25G TCD4004-5G
1,1-Dibromo-3,3,3-trifluoroacetone ≥98.0% (by GC)
1,1-Dibromo-3,3,3-trifluoroacetone
Formula: Br₂HCC(O)CF₃
MW: 269.84 g/mol
Boiling Pt: 113 °C
MDL Number: MFCD00041362
CAS Number: 431-67-4
UN: 3265
ADR: 8 II

Order Now

Specification Test Results

Appearance Colorless to Red to Green clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR