1-Bromo-4-fluorobutane ≥98.0% (by GC)

Supplier: TCI America
Warning

B5331-25G B5331-5G
TCB5331-25GEA 157 USD
TCB5331-25G TCB5331-5G
1-Bromo-4-fluorobutane ≥98.0% (by GC)
1-Bromo-4-fluorobutane
Formula: BrH₂C(CH₂)₂CH₂F
MW: 155.01 g/mol
Boiling Pt: 135 °C
Flash Pt: 45 °C
MDL Number: MFCD00209551
CAS Number: 462-72-6
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR