8-Chloro-1-(3-chloropropyl)theobromine ≥95.0% (by HPLC)
Supplier: TCI America
Synonyms:
8-Chloro-1-(3-chloropropyl)-3,7-dimethyl-1H-purine-2,6(3H,7H)-dione
C3618-25G
C3618-5G
TCC3618-25GEA
403
USD
TCC3618-25G
TCC3618-5G
8-Chloro-1-(3-chloropropyl)theobromine ≥95.0% (by HPLC)
8-Chloro-1-(3-chloropropyl)theobromine
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Melting point | 144.0 to 148.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...