Cyclohexanemethylamine hydrobromide ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: (Aminomethyl)cyclohexane hydrobromide, Cyclohexylmethylamine hydrobromide, Cyclohexanemethylammonium bromide

C3531-1G C3531-5G
TCC3531-1GEA 46 USD
TCC3531-1G TCC3531-5G
Cyclohexanemethylamine hydrobromide ≥98.0% (by titrimetric analysis)
Cyclohexanemethylamine hydrobromide

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR