AMI-331 hydrochloride ≥97.0% (by HPLC)
Supplier: TCI America
Synonyms:
3-Bromo-1-(prop-2-yn-1-yl)-2-(pyridin-4-yl)-1,5,6,7-tetrahydro-4H-pyrrolo[3,2-c]pyridin-4-one hydrochloride
A3352-25MG
A3352-5MG
TCA3352-25MGEA
727
USD
TCA3352-25MG
TCA3352-5MG
AMI-331 hydrochloride ≥97.0% (by HPLC)
AMI-331 hydrochloride
Specification Test Results
Appearance | White to Amber powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...