4,5-Bis(dicyclohexylphosphino)-9,9-dimethylxanthene ≥97.0% (by GC)
Supplier: TCI America
Synonyms:
4,5-Bis(dicyclohexylphosphino)-9,9-dimethyl-9H-xanthene
B5239-1G
B5239-200MG
TCB5239-1GEA
159
USD
TCB5239-1G
TCB5239-200MG
4,5-Bis(dicyclohexylphosphino)-9,9-dimethylxanthene ≥97.0% (by GC)
4,5-Bis(dicyclohexylphosphino)-9,9-dimethylxanthene
Formula:
C₃₉H₅₆OP₂ MW: 602.82 g/mol |
MDL Number:
MFCD22422615 CAS Number: 940934-47-4 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 169.0 to 174.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...