2-N-Propyl-4-methyl-6-(1'-methylbenzimidazole-2-yl)benzimidazole ≥98.0% (by HPLC)

Supplier: TCI America

D5079-25G D5079-5G
TCD5079-25GEA 329 USD
TCD5079-25G TCD5079-5G
2-N-Propyl-4-methyl-6-(1'-methylbenzimidazole-2-yl)benzimidazole ≥98.0% (by HPLC)
2-N-Propyl-4-methyl-6-(1'-methylbenzimidazole-2-yl)benzimidazole
Formula: C₁₉H₂₀N₄
MDL Number: MFCD09029193
CAS Number: 152628-02-9

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystaline
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR