cis-N-(tert-Butoxycarbonyl)-4-hydroxy-D-proline ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: Boc-cis-D-Hyp-OH, (2R,4R)-1-(tert-Butoxycarbonyl)-4-hydroxypyrrolidine-2-carboxylic acid, (2R,4R)-N-Boc-4-hydroxy-2-pyrrolidinecarboxylic acid

B5886-1G B5886-5G
TCB5886-1GEA 32 USD
TCB5886-1G TCB5886-5G
cis-N-(tert-Butoxycarbonyl)-4-hydroxy-D-proline ≥98.0% (by HPLC, titration analysis)
cis-N-(tert-Butoxycarbonyl)-4-hydroxy-D-proline
Formula: C₁₀H₁₇NO₅
MW: 231.25 g/mol
MDL Number: MFCD02094407
CAS Number: 135042-12-5

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D +46.0 to +54.0 deg (C=1, MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR