4-[[4,6-Bis(n-octylthio)-1,3,5-triazin-2-yl]amino]-2,6-di-tert-butylphenol ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

B5977-25G B5977-5G
TCB5977-25GEA 223 USD
TCB5977-25G TCB5977-5G
4-[[4,6-Bis(n-octylthio)-1,3,5-triazin-2-yl]amino]-2,6-di-tert-butylphenol ≥98.0% (by HPLC, total nitrogen)
4-[[4,6-Bis(n-octylthio)-1,3,5-triazin-2-yl]amino]-2,6-di-tert-butylphenol
Formula: C₃₃H₅₆N₄OS₂
MDL Number: MFCD00134696
CAS Number: 991-84-4
EINECS: 213-590-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 93.0 to 97.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR