3-Bromodibenzothiophene ≥98.0% (by GC)

Supplier: TCI America

B5452-1G B5452-5G
TCB5452-1GEA 134 USD
TCB5452-1G TCB5452-5G
3-Bromodibenzothiophene ≥98.0% (by GC)
3-Bromodibenzothiophene
Formula: C₁₂H₇BrS
MDL Number: MFCD11052997
CAS Number: 97511-04-1

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 97.0 to 101.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR