Brequinar ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 6-Fluoro-2-(2'-fluoro-[1,1'-biphenyl]-4-yl)-3-methylquinoline-4-carboxylic acid

B5707-100MG B5707-20MG
TCB5707-100MGEA 532 USD
TCB5707-100MG TCB5707-20MG
Brequinar ≥98.0% (by HPLC)
Brequinar
Formula: C₂₃H₁₅F₂NO₂
MW: 375.37 g/mol
Melting Pt: 317 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00866437
CAS Number: 96187-53-0

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 3.50 to 4.00 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR