3,3''-Dibromo-1,1':3',1''-terphenyl ≥97.0% (by GC)

Supplier: TCI America

D4999-1G D4999-200MG
TCD4999-1GEA 149 USD
TCD4999-1G TCD4999-200MG
3,3''-Dibromo-1,1':3',1''-terphenyl ≥97.0% (by GC)
3,3''-Dibromo-1,1':3',1''-terphenyl
Formula: C₁₈H₁₂Br₂
Boiling Pt: 225 °C
MDL Number: MFCD28138089
CAS Number: 95962-62-2
UN: 3152
ADR: 9,II

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR