9-(4-Bromophenyl)-10-(2-naphthyl)anthracene ≥97.0% (by HPLC)

Supplier: TCI America

B5030-1G B5030-200MG
TCB5030-1GEA 303 USD
TCB5030-1G TCB5030-200MG
9-(4-Bromophenyl)-10-(2-naphthyl)anthracene ≥97.0% (by HPLC)
9-(4-Bromophenyl)-10-(2-naphthyl)anthracene
Formula: C₃₀H₁₉Br
MW: 459.39 g/mol
MDL Number: MFCD26407508
CAS Number: 936854-62-5

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Argentmetric Titration) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR