[3,5-Bis(ethoxycarbonyl)phenyl]boronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

B6032-1G B6032-200MG
TCB6032-1GEA 120 USD
TCB6032-1G TCB6032-200MG
[3,5-Bis(ethoxycarbonyl)phenyl]boronic acid (contains varying amounts of Anhydride)
[3,5-Bis(ethoxycarbonyl)phenyl]boronic acid
Formula: C₁₂H₁₅BO₆
MDL Number: MFCD11867795
CAS Number: 932378-94-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 and le 107.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR