Chloro[[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene](N,N-dimethylbenzylamine)palladium(II)] ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: SingaCycle(TM)-A1

C2372-1G C2372-200MG
TCC2372-1GEA 646 USD
TCC2372-1G TCC2372-200MG
Chloro[[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene](N,N-dimethylbenzylamine)palladium(II)] ≥97.0% (by titrimetric analysis)
Chloro[[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene](N,N-dimethylbenzylamine)palladium(II)]

This product is unavailable in the U.S.

Formula: C₃₆H₄₈ClN₃Pd
MW: 664.67 g/mol
MDL Number: MFCD15072151
CAS Number: 930796-10-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Chelometric Titration) min. 97.0 %
Purity (with Total Nitrogen) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR