Acridine-3,6-diamine ≥96.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 3,6-Diaminoacridine, Proflavine, 3,6-Acridinediamine

A3219-1G A3219-5G
TCA3219-1GEA 124 USD
TCA3219-1G TCA3219-5G
Acridine-3,6-diamine ≥96.0% (by HPLC, titration analysis)
Acridine-3,6-diamine
Formula: C₁₃H₁₁N₃
MDL Number: MFCD00005030
CAS Number: 92-62-6
EINECS: 202-172-4
Merck Index: 14,07771

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 96.0 area%
Purity (Nonaqueous Titration) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR