2-Bromo-5-(2-ethylhexyl)thiophene ≥98.0% (by GC)

Supplier: TCI America

B5475-1G B5475-5G
TCB5475-1GEA 162 USD
TCB5475-1G TCB5475-5G
2-Bromo-5-(2-ethylhexyl)thiophene ≥98.0% (by GC)
2-Bromo-5-(2-ethylhexyl)thiophene
Formula: C₁₂H₁₉BrS
MW: 275.25 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28101692
CAS Number: 925899-21-4

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR