9,9-Bis[4-[di(2-naphthyl)amino]phenyl]fluorene ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

B4978-1G B4978-200MG
TCB4978-1GEA 501 USD
TCB4978-1G TCB4978-200MG
9,9-Bis[4-[di(2-naphthyl)amino]phenyl]fluorene ≥98.0% (by HPLC, total nitrogen)
9,9-Bis[4-[di(2-naphthyl)amino]phenyl]fluorene
Formula: C₆₅H₄₄N₂
MW: 853.08 g/mol
CAS Number: 916061-87-5

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR