2-([1,1'-Biphenyl]-3-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)biphenyl, 3-Biphenylboronic acid Pinacol ester

B6023-1G B6023-5G
TCB6023-1GEA 38 USD
TCB6023-1G TCB6023-5G
2-([1,1'-Biphenyl]-3-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥98.0% (by GC, titration analysis)
2-([1,1'-Biphenyl]-3-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formula: C₁₈H₂₁BO₂
MW: 280.17 g/mol
MDL Number: MFCD18452169
CAS Number: 912844-88-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 80.0 to 84.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR