2,8-Bis(diphenylphosphoryl)dibenzo[b,d]furan ≥96.0% (by HPLC)
Supplier: TCI America
Synonyms:
Dibenzo[b,d]furan-2,8-diylbis(diphenylphosphine oxide)
B5822-1G
B5822-200MG
TCB5822-1GEA
402
USD
TCB5822-1G
TCB5822-200MG
2,8-Bis(diphenylphosphoryl)dibenzo[b,d]furan ≥96.0% (by HPLC)
2,8-Bis(diphenylphosphoryl)dibenzo[b,d]furan
Formula:
C₃₆H₂₆O₃P₂ MW: 568.55 g/mol |
MDL Number:
MFCD30607231 CAS Number: 911397-27-8 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 96.0 area% |
Melting point | 251.0 to 255.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...