6,7-Bis(benzyloxy)coumarin ≥98.0% (by HPLC)

Supplier: TCI America

B5251-1G B5251-200MG
TCB5251-1GEA 199 USD
TCB5251-1G TCB5251-200MG
6,7-Bis(benzyloxy)coumarin ≥98.0% (by HPLC)
6,7-Bis(benzyloxy)coumarin
Formula: C₂₃H₁₈O₄
MW: 358.39 g/mol
MDL Number: MFCD00016975
CAS Number: 909-84-2
EINECS: 213-003-9

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 161.0 to 165.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR