N,N-Bis[(diphenylphosphino)methyl]-3-(triethoxysilyl)propylamine ≥95.0% (by total nitrogen basis)

Supplier: TCI America
Warning

Synonyms: 3-[Bis[(diphenylphosphino)methyl]amino]propyltriethoxysilane

B5594-1G B5594-5G
TCB5594-1GEA 101 USD
TCB5594-1G TCB5594-5G
N,N-Bis[(diphenylphosphino)methyl]-3-(triethoxysilyl)propylamine ≥95.0% (by total nitrogen basis)
N,N-Bis[(diphenylphosphino)methyl]-3-(triethoxysilyl)propylamine
Formula: C₃₅H₄₅NO₃P₂Si
MW: 617.78 g/mol
Storage Temperature: Refrigerator
CAS Number: 904704-23-0

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (with Total Nitrogen) 95.0 to 105.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR