N-[4-Cyano-3-(trifluoromethyl)phenyl]-2-methyloxirane-2-carboxamide ≥95.0% (by GC)

Supplier: TCI America
Warning

C3607-25G C3607-5G
TCC3607-25GEA 131 USD
TCC3607-25G TCC3607-5G
N-[4-Cyano-3-(trifluoromethyl)phenyl]-2-methyloxirane-2-carboxamide ≥95.0% (by GC)
N-[4-Cyano-3-(trifluoromethyl)phenyl]-2-methyloxirane-2-carboxamide
Formula: C₁₂H₉F₃N₂O₂
MDL Number: MFCD08460201
CAS Number: 90357-51-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Melting point 151.0 to 155.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR